Whatman™ Filters for Water, Air and Soil Testing
Accurate and reliable analytical results are important when monitoring air, water and soil for Environmental testing. GE knows that you need to be sure your filtration step is efficient, reproducible and preserves the integrity of your sample. Whatever you are using one of our glass papers, syringe filters or other Whatman™ products, you can count on GE to understand that quality matters.
Water Testing   Air Testing   Soil Testing
NEW RTU* filters available!
*Ready-To-Use
40 vuotta ympäristönäytteiden valmistelu- ja testauskokemusta (kesto 21 minuuttia)
Top
Whatman™ Filters for Water Testing
With Whatman™ filters you can test your samples according to the environmental water testing requirements. The following testing have to be performed:
  • solids analysis
  • metals analysis
  • dissolution ions sample preparation
  • dissolved organic carbons content
  • HPLC, UHPL, ICP-MS
  • microbiological analysis
Whatman™ has developed specific filters to guarantee reliable results and to let you perform easily each analysis even starting from critical samples.
For example, with the 934-AH RTU glass fiber filters, which are supplied in a prewashed and preweighed format,  enable a considerable time saving when performing suspended solid analysis.
The GD/XP syringe filters, including an integrated prefiltration with dual-layer prefilter stack and one final 0.45 micron membrane, allow an easy filtration of hard-to-filter samples. Futhermore the pre-filter in polypropylene minimizes ion extractables making them suitable for metal trace analysis by ICP-MS. Find out how Anotop™ IC, Puradisc Aqua 30, Puradisc, Spartan, GD/XP and GD/X™ and Mini-Uniprep can help you!
Customer testimonial: Whatman™ filters for Water Quality Testing
Uutuus: RTU (Ready-To-Use) -tuotesarjaan on nyt lisätty myös suodattimet
GF/C RTU liuenneille ja suspendoituneille kiintoaineille standardin EN872 mukaisesti
934-AH RTU helposti haihtuville aineille US EPA -laboratoriostandardin menetelmän 2540 C ja D mukaisesti
Käyttövalmiit: esipesty, kuivattu/poltettu, jäähdytetty, vesi poistettu, punnittu 4 desimaalin tarkkuudella, asetettu merkittyihin/viivakoodillisiin alustoihin ja pakattu viivakoodilliseen laatikkoon 
Physical Analysis
Total suspended, dissolved and volatile solids have to be determined by using glass fibre filters. The 934-AH RTU glass fiber filters are ready for use!
Glass microfibre filters without binder, grade GF/C, Whatman
Application:
Filters for total suspended, dissolved and volatile solids analysis
Highlight 1:
Retention of very fine particles
Highlight 2:
Particle rentention 1,2 µm
Standard Method Compliance:
EN 872
Filter name:
GF/C
Filter type:
Glass fiber filters
Glass microfibre filters without binder, grade 934-AH, Whatman
Application:
Filters for total suspended, solids analysis
Highlight 1:
High loading capacity
Standard Method Compliance:
2540D
Filter name: 934-AH
Filter Type: Glass fiber filter
Nyt myynnissä uudet RTU-tuotteet haihtuville aineille
Glass microfibre filters, 934-AH™ RTU, Whatman
Application:
Filters for total suspended, solids analysis
Highlight 1:
Ready for use: prewashed and preweighed
Highlight 2:
Each pan with unique barcode
Standard Method Compliance:
2540D
Filter name: 934-AH RTU
Filter Type: Glass fiber filter
Glass microfibre filters without binder, grade GF/C RTU, Whatman
Application:
Filters for total suspended, dissolved and volatile solids analysis
Highlight 1:
Ready for use: prewashed, dried, cooled and weighed
Highlight 2:
Certified mass loss <0.017 mg/cm2 
Standard Method Compliance:
EN 872
Filter name:GF/C RTU
Filter Type: Glass fiber filter
Demo of Glass microfiber filters, 934-AH™ RTU, Whatman
Chemical Analysis
Chemical analysis are commonly performed using analytical instrumentation. Filtration of water samples prior to analyis is good practice in order to remove unwanted particles from the analysis and to protect delicate instrumentation from potentially damaging compounds.
In this section you can find filters for:
  • dissolved heavy metals determination (GD/XP™ Syringe filters, In-line filters Polydisc™ GW and Polycap™ GW)
  • dissolution ions sample preparation (Anotop™ IC syringe filter)
  • dissolved organic carbon (DOC) determination (Puradisc™ Aqua 30 syringe filter )
Syringe Filters, GD/XP™, Whatman
Application:
Dissolved heavy metals analysis
Highlight 1:
For hard-to filter samples
Highlight 2:
Suitable for metal trace analysis
Filter name:
GD/XP™
Filter Type:
SYRINGE FILTERS
In-Line Filters, Polydisc™ GW, Whatman
Application:
Dissolved heavy metals analysis
Highlight 1:
For ground water samples
Filter name:
POLYDISC™ GW
Filter Type:
In-Line FILTERS
Groundwater Sampling Capsules, Polycap™ GW, Whatman
Application:
Dissolved heavy metals analysis
Highlight 1:
For ground water samples
Filter name:
POLYCAP™ GW
Filter Type:
In-Line FILTERS
Syringe Filters, Anotop™, Whatman
Application:
Dissolved ions analysis
Highlight 1:
Certified and guaranteed low levels of anion leaching
Filter name:
ANOTOP™ IC
Filter Type:
SYRINGE FILTERS
Syringe Filters, Puradisc™ Aqua 30, Whatman
Application:
Dissolved organic carbon analysis
Highlight 1:
Prewashed
Filter name:
PURADISC™ AQUA30
Filter Type:
SYRINGE FILTERS
HPLC/UHPLC and Other Analytical Techniques
Microbiological Analysis
Related Items
VWR can provide you a complete product range of consumables, equipments and reagents for Environmental analysis. Use the search tool in our home page or contact your sales representative.
Contact
Top
Whatman™ Filters for Air Testing
Air quality monitoring is getting more and more important because of the presence of high amount of air pollutants in the ambient air. The sources of emission can be natural or caused by industries, agriculture or transport traffic. Ultrafine particles (UFPs) have become a key area of interest in environmental monitoring and related studies as they have been shown to have potentially significant impacts on air quality and health.
Heavy metals, organics and inorganics chemicals such as ozone, volatile organic carbons, SO2, NO2, CO, benzoate, or asbestos has to be monitored too.
Whatman™ can support you with dedicated filters for particulate monitoring both manual and automated; for sampling in normal or aggressive enviroments; for radioactivity, metals or asbestos testing.
Make your choice between 13 different types of Whatman™ filters for air sample testing:
  • PM2,5
  • GF/A
  • EPM 2000
  • GF10
  • TE membranes
  • QM-A quartz fiber filters
  • QM-H quartz fiber filters
  • QM-A quartz thimbles
  • QM-H quartz thimbles
  • Glass microfiber reels
  • WME
  • Nucleopore™ membranes
  • Cyclopore™ membranes
Particulate Analysis
Manual sampling in normal environments
Glass Microfibre Filters with Binder, Grade GF 10, Whatman
Application:
Manual sampling of ambient air particulate (normal environment)
Highlight 1:
Extreme mechanical stability
Highlight 2:
Used up to 180 °C
Filter name:
GF10
Filter Type:
GLASS FIBER FILTERS
Extraction thimbles, Whatman
Application:
Manual sampling of ambient air particulate (normal environment)
Highlight 1:
For stack gases
Highlight 2:
Used up to 500 °C
Name filter:
GLASS FIBRE THIMBLES
Filter Type:
GLASS FIBRE THIMBLES
Membrane Filters, PTFE, TE Range, Whatman
Application:
Manual sampling of ambient air particulate (normal environment)
Highlight 1:
High chemical compatibility
Highlight 2:
Low chemical background interference
Filter name:
TE PTFE
Filter Type:
PTFE MEMBRANE FILTERS
Glass Microfibre Filters without Binder, Grade GF/A, Whatman
Application:
Manual sampling of ambient air particulate (normal environment)
Highlight 1:
For gravimetric determination, PM10
Highlight 2:
High-flow rate
Filter name:
GF/A
Filter Type:
GLASS FIBER FILTERS
Glass Microfibre Filters for Air Sampling, Grade EPM 2000, Whatman
Application:
Manual sampling of ambient air particulate (normal environment)
Highlight 1:
For gravimetric determination, PM10
Highlight 2:
Used in high-volume PM-10 air sampling equipment
Name filter:
EPM2000
Filter Type:
GLASS FIBER FILTERS
Membrane filters for air sampling, PTFE, Grade PM 2,5, Whatman
Application:
Manual sampling of ambient air particulate (normal environment)
Highlight 1:
For PM 2.5 determination. High chemical compatibility
Highlight 2:
Low chemical background interference
Standard Method compliance:
EPA PM 2.5 (requirements of 40 CFR Part 50 Appendix L)
Filter name:
PM 2.5 PTFE
Filter Type:
PTFE MEMBRANE FILTERS
Manual sampling in aggressive environments
Quartz Microfibre Filters for Air Sampling, Grade QM-A, Whatman
Application:
Manual sampling of ambient air particulate (aggressive environment)
Highlight 1:
For stack gases. High-purity quartz.
Highlight 2:
Used up to 800 °C
Filter name:
QM-A
Filter Type:
QUARTZ FIBER FILTERS
Pure Quartz Air Sampling Filters, Grade QM-H, Whatman
Application:
Manual sampling of ambient air particulate (aggressive environment)
Highlight 1:
100% pure quartz
Highlight 2:
Used up to 900 °C
Type of filter:
QM-H
Filter Type:
QUARTZ FIBER FILTERS
Extraction Thimbles, High Purity, Quartz, Whatman
Application:
Manual sampling of ambient air particulate (aggressive environment)
Highlight 1:
High-purity quartz. For stack gases.
Highlight 2:
Used up to 800 °C
Filter name:
QUARTZ FIBER THIMBLES
Filter Type:
QUARTZ FIBER THIMBLES
Automated sampling
Glass Microfibre Filters with Binder, Grade GF 10, Whatman
Application:
Automated sampling of ambient air particulate
Highlight 1:
Extreme mechanical stability
Highlight 2:
Used up to 180 °C
Filter name:
GF10 REELS
Filter Type:
GLASS MICROFIBER REELS
Sampling for radioactivity and metal analysis
Glass Microfibre Filters without Binder, Grade GF/A, Whatman
Application:
Sampling of radioactive air pollution
Highlight 1:
Suitable to absorb radioactive iodine
Highlight 2:
High-flow rate
Filter name:
GF/A
Filter Type:
GLASS FIBER FILTERS
Application Specific Filter Papers, Grade 72, Whatman
Application:
Sampling of radioactive air pollution
Highlight 1:
Suitable to absorb radioactive iodine
Filter name :
Grade 72
Filter Type:
GLASS FIBRE/CELLULOSE FILTER PAPER
Filters for the determination of metals in ambient particulate
Membrane Filters, Mixed Cellulose Ester, Whatman
Application:
Filters for the determination of metals in ambient particulate
Highlight 1:
Good-flow rate and strenght
Highlight 2:
Wide range of porosities
Filter name:
MCE
Filter Type:
MCE MEMBRANE FILTERS
Chemical Analysis
Air pollution monitoring from stacks, flues and aerosols requires a filter that can whithstand chemically harsh environments and high temperatures. High-purity is also required for this application.
Filters for heavy metals analysis
Quartz Microfibre Filters for Air Sampling, Grade QM-A, Whatman
Application:
Heavy metals analysis
Highlight 1:
High-purity quartz
Highlight 2:
Used up to 800 °C
Filter name:
QM-A
Filter Type:
QUARTZ FIBER FILTERS
Pure Quartz Air Sampling Filters, Grade QM-H, Whatman
Application:
Heavy metals analysis
Highlight 1:
100% pure quartz
Highlight 2:
Used up to 900 °C
Type of filter:
QM-H
Filter Type:
QUARTZ FIBER FILTERS
Glass Microfibre Filters for Air Sampling, Grade EPM 2000, Whatman
Application:
Heavy metals analysis
Highlight 1:
Detailed chemical analysis of trace pollutants
Highlight 2:
Used in high-volume PM-10 air sampling equipment
Filter name:
EPM2000
Filter Type:
GLASS FIBER FILTERS
Extraction Thimbles, High Purity, Quartz, Whatman
Application:
Heavy metals analysis
Highlight 1:
High-purity quartz. Suitable for both solvent extraction and air sampling.
Highlight 2:
Used up to 800 °C
Filter name:
QUARTZ FIBER THIMBLES
Filter Type:
QUARTZ FIBER THIMBLES
Filters for other organic or inorganic chemicals
Glass Microfibre Filters without Binder, Grade GF/A, Whatman
Application:
Other organic or inorganic chemicals
Highlight 1:
High-flow rate
Filter name:
GF/A
Filter Type:
GLASS FIBER FILTERS
Membrane Filters, PTFE, TE Range, Whatman
Application:
Other organic or inorganic chemicals
Highlight 1:
Increased durability for aggressive testing envirnments
Highlight 2:
High chemical compatibility
Type of filter:
TE PTFE
Filter Type:
PTFE MEMBRANE FILTERS
Quartz Microfibre Filters for Air Sampling, Grade QM-A, Whatman
Application:
Other organic or inorganic chemicals
Highlight 1:
High-purity quartz
Highlight 2:
Used up to 800 °C
Filter name:
QM-A
Filter Type:
QUARTZ FIBER FILTERS
Pure Quartz Air Sampling Filters, Grade QM-H, Whatman
Application:
Other organic or inorganic chemicals
Highlight 1:
100% pure quartz
Highlight 2:
Used up to 900 °C
Filter name:
QM-H
Filter Type:
QUARTZ FIBER FILTERS
Extraction Thimbles, High Purity, Quartz, Whatman
Application:
Other organic or inorganic chemicals
Highlight 1:
High-purity quartz. Suitable for both solvent extraction and air sampling.
Highlight 2:
Used up to 800 °C
Type of filter:
QUARTZ FIBER THIMBLES
Filter Type:
QUARTZ FIBER THIMBLES
Asbestos Analysis
Related Items
VWR can provide you a complete product range of consumables, equipments and reagents for Environmental analysis. Use the search tool in our home page or contact your sales representative.
Contact
Top
Whatman™ Filters for Soil Testing
From an environmental point of view, soil is a primary element of the ecosystem, together with water, air and living organisms, and therefore needs to be protected and preserved by possible degradation phenomena. On the other hand, the use of fertilizers for agricultural activity is a powerful means of increasing the productivity of the soils and therefore yields of crops.
Soil sample analysis requires that chemical compounds are first extracted from their matrix using pH and heat modulation and concentrated. Nitrogen, pesticides, phosporous, retained solids and clarified solutions, pH have to be determined in soil samples. Whatman™ can provide you the right product for each testing.
Request Whatman filter samples for SOIL
Kjeldal nitrogen analysis
Kjeldal nitrogen analysis
Soil nitrogen content is typically done with Kjeldal techniques, which involve the sampling of an exact amount of soil before transfer to a digestion tube. For this aplication you need to use the Whatman™ low nitrogen content weighing boats and then filter the sample with qualitative paper.
Qualitative Filter Papers, Standard Grades, Grade 4 and 4V, Whatman
Application:
Kjeldal nitrogen analysis
Highlight 1:
For sample filtration prior to analysis
Filter name:
Grade 4
Filter Type:
FILTER PAPER
Kjeldahl Weighing Boats, Whatman
Application:
Kjeldal nitrogen analysis
Highlight 1:
Low nitrogen content
Type of filter:
Grade 609
Filter Type:
WEIGHING BOATS
Soxhlet Extraction
Prior to analysis by, for example, gas chromatography (GC), soils may be prepared using Soxhlet extraction or microwave digestion. Extraction thimbles are widely used for this technique. To remove fine particles and protect your GC instrument use specific syringe filters.
Extraction Thimbles, Whatman
Application:
Soxhlet extraction
Filter name: CELL THIMBLES
Filter Type: CELLULOSE THIMBLES
Spectrophotometry and Chromatography
Most trace element tests are based on extracting soil and measuring the concentration of trace elements in the soil-free liquid phase using for example inductively coupled plasma atomic emission spectrometry (ICP-AES). The sample then generally needs to be filtered through a qualitative filter paper or glass fiber filter to make sure it will not clog nebulizers or interfere with injection into the analysis instrument. If digested with acqua regia, the sample may be filtered through an ashless filter paper.
Syringe Filters, SPARTAN™, Whatman
Application:
Spectrophotometry and Chromatography
Highlight 1:
HPLC certified
Filter name:
SPARTAN
Filter Type:
Glass fiber filter
Syringe Filters, Puradisc™, Whatman
Application:
Spectrophotometry and Chromatography
Highlight 1:
Wide range of membranes, pore sizes and diameters
Filter name:PURADISC
Filter Type:Glass fiber filter
Syringe Filters, GD/X™, Whatman
Application:
Spectrophotometry and Chromatography
Highlight 1:
With prefiltration stack of glass fiber filter
Highlight 2:
Three to seven times more sample volume than standard syringe filters
Filter name:
GD/X
Filter Type:
Syringe filters
Syringe Filters, GD/XP™, Whatman
Application:
Hard-to-filter samples
Highlight 1:
With polypropylene prefilter stack
Highlight 2:
Minimize the levels of ion extractables
Filter name:
GD/XP
Filter Type:
Syringe filters
Syringeless Filters, Mini-UniPrep™ G2, Whatman
Application:
Spectrophotometry and Chromatography
Highlight 1:
All-in-one filter and autosampler vial
Highlight 2:
With GLASS autosampler vial
Filter name:
MINIUNIPREP G2
Filter Type:
Glass fiber filter
Syringeless Filters, Mini-UniPrep™, Whatman
Application:
Spectrophotometry and Chromatography
Highlight 1:
All-in-one filter and autosampler vial
Highlight 2:
With PLASTIC autosampler vial
Filter name:
MINIUNIPREP
Filter Type:
Glass fiber filter
Demo of Whatman GD/X™ syringe filter
Demo of Whatman Mini-UniPrep™
Colorimetry for Phosphorous Analysis
Through soil phosphorus testing, the amount of phosphorus fertilizer required to achieve maximum plant growth can be determined. To determine the soil phosphorus content, the soil is extracted with a chemical solution and the phosphorus content in the extract is measured by colorimetry. Filtration of the extract through a qualitative filter paper is generally needed before analysis. If an automated method is used for determining phosphorus concentration, acid-resistant filter paper may be needed.
Quantitative Filter Papers, Ashless Grades, Grade 41, Whatman
Application:
Colorimetry for phosphorous analysis
Filter name: Grade 41
Filter Type: FILTER PAPER
pH analysis
The pH of soil is vital for how well it holds minerals. Some plants have very particular pH requirements. There are many different ways of measuring soil pH. Litmus/pH paper is a quick and inexpensive method to test soil pH when a pH-meter is unavailable or when highly precise values are not necessary. When preparing your soil sample, use a weighing paper to weigh the soil before adding water. Filter papers can be used to remove unnecessary particles from the suspension.
pH indicator papers, Whatman
Application:
pH Analysis
Highlight 2:
Universal indicators
Filter name:
PHANPEHA
Filter Type:
INDICATOR PAPERS
Whatman™ filters for sample clarification and solids retention
Various test methods require that liquid components of a solution be separated from suspended solids prior to analysis. GE offers a wide choice of cellulose filter papers with different levels of flow rate, loading capacity, and chemical resistance to support these applications.
Paper Filters: Selector Guide
Related Items
VWR can provide you a complete product range of consumables, equipments and reagents for Environmental analysis. Use the search tool in our home page or contact your sales representative.
Contact