Cavity slide
Toimittaja: Karl Hecht
HECH42432010EA 0 EUR
HECH42432010
Cavity slide
This microscopy slide is made of opal glass.
  • Opal 'milk' glass
  • Frosted label
  • Marking
Suitable for blood typing.