Revco high performance laboratory refrigerators
Toimittaja: Thermo Scientific
High performance laboratory refrigerator stores samples for clinical, industrial and scientific applications.
  • Digital electronic technology designed to simplify operation and deliver greater accuracy
  • Large, easy-to-read digital display of temperature
  • Controller battery backup system
  • High density CFC-free urethane foam insulation
These refrigerators feature a microprocessor control system, positive airflow systems, industrial-quality cabinet construction and extra-strength refrigeration compressors.
Tilaustiedot: Alarm systems, temperature chart recorders and power monitors to be ordered separately.
TILAUS
Laajenna kaikki / Pienennä kaikki
Tyyppi Pak. VWR-tuoteluettelon nro Määrä
General purpose laboratory refrigerators, space saver, REL-1204V 1 REVCREL1204V
General purpose laboratory refrigerators, upright, REL-2304V 1 REVCREL2304V
General purpose laboratory refrigerators, upright, REL-3004V 1 REVCREL3004V
General purpose laboratory refrigerators, undercounter, REL-404V 1 REVCREL404V
General purpose laboratory refrigerators, upright, REL-4504V 1 REVCREL4504V
General purpose laboratory refrigerators, upright, REL-5004V 1 REVCREL5004V
General purpose laboratory refrigerators, upright, REL-7504V 1 REVCREL7504V
General purpose laboratory refrigerators, upright, RGL5004V 1 REVCRGL5004V
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
TEKNISET TIEDOT
Tyyppi REL-7504V REL-1204V REL-5004V REL-404V RGL-5004V REL-2304V REL-4504V REL-3004V
Tilavuus (l) 2231 326 1447 133 1447 659 1297 826
Lämpötila (°C) +1...+8
Ilmankierto Puhallin
Näyttö digital
Sulatus automatic
Hälytys audible and visual
Hyllyt/laatikot 12 4 8 3 8 4 8 4
Ovi/ovet 3 solid doors 1 solid door 2 solid doors 1 solid door 2 glass doors 1 solid door 2 sliding glass doors 1 solid door
Ovilukko yes
Materiaali Terästä, jauhepinnoitettu
Läpivientiaukot 1
L×S×K sisämitat (mm) 2050×730×1470 500×530×1320 1320×730×1470 500×500×500 1334×737×1473 610×730×1470 1320×660×1470 760×730×1470
L×S×K ulkomitat (mm) 2160×910×1970 610×740×1830 1440×910×1970 610×660×850 1435×945×2012 720×910×1970 1440×890×1970 870×910×1970
Paino (kg) 365 127 265 96 265 184 304 197
Syötä tarkemmat tiedot.
Valitse tuote johon etsit Sertifikaattia.
Ilmoita eränumero. Tämä kenttä on pakollinen. Jos et tiedä VWR Chemicalsin eränumeroa syötä "x".
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
tai
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Kuvaus Tuoteluettelon nro Availability Tuote Sinun hintasi Price Per lkm