SPE columns, CHROMABOND® C18 Hydra
Toimittaja: Macherey-Nagel
CHROMABOND®
MANA730301EA 451 EUR
MANA730301 MANA730300 MANA730299 MANA730297 MANA730296 MANA732295 MANA738294.100M
SPE columns, CHROMABOND® C18 Hydra
Kromatografiakolonnit
CHROMABOND® C18 Hydra is a special octadecyl modified silica phase for more polar analytes in SPE.
  • Slightly polar
  • Hydrophobic and strong secondary interactions with polar groups of the analytes
  • Suitable for nonpolar and polar substances from water
Recommended for more polar compounds like pesticides and their polar degradation products, phenols, phenoxycarboxylic acids, nitroaromatics and pharmaceuticals.