VWR® Rapid Formaldehyde-free RNA Gel Kit
Toimittaja: Avantor
VWR®
1B1384-KITEA 356 EUR
1B1384-KIT
VWR® Rapid Formaldehyde-free RNA Gel Kit
Elektroforeesigeelit Agaroosi ja akryyliamidi
The Rapid formaldehyde-free RNA gel kit is a safer alternative to formaldehyde-containing agarose gels for denaturing electrophoresis of RNA. The kit provides a non-volatile substitute for formaldehyde as well as a non-mutagenic RNA stain, which eliminates the need for ethidium bromide staining.
  • Hood-free pouring and running - completely formaldehyde-free
  • Denature and stain samples in a single step
  • Instant band visualisation
  • Compatible with down-stream applications, including Northern Blots
The non-volatile denaturing agent included in the gel solution and loading buffer of both kits effectively eliminates RNA secondary structure to ensure optimal resolution during electrophoresis. Gel casting and electrophoresis can be safely performed on the benchtop without a fume hood. In contrast to formaldehyde-free gel kits containing glyoxal, this kit does not require extended incubation steps or buffer recirculation.
  • Hood-free pouring and running - completely formaldehyde-free
  • Denature and stain samples in a single step
  • Instant band visualisation
  • Compatible with down-stream applications, including Northern Blots
The non-volatile denaturing agent included in the gel solution and loading buffer of both kits effectively eliminates RNA secondary structure to ensure optimal resolution during electrophoresis. Gel casting and electrophoresis can be safely performed on the benchtop without a fume hood. In contrast to formaldehyde-free gel kits containing glyoxal, this kit does not require extended incubation steps or buffer recirculation.
Toimitustiedot: Kit includes Rapid RNA gel running buffer, 10X (2×500 ml), Rapid formaldehyde-free RNA gel solution, 10X (150 ml), and formaldehyde-free RNA gel loading buffer, 2X (3×2 ml). Each kit contains sufficient material for 30×50 ml gels.