3-Hydroxy-1,8-naphthalic anhydride ≥98.0% (by titrimetric analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAH0278-5GEA 377 EUR
TCIAH0278-5G
3-Hydroxy-1,8-naphthalic anhydride ≥98.0% (by titrimetric analysis)
3-Hydroxy-1,8-naphthalic anhydride
Kaava: C₁₂H₆O₄
MW: 214,17 g/mol
MDL numero: MFCD00059759
CAS-numero: 23204-36-6

Order Now

Testitulosten erittely

Appearance Light yellow to Yellow powder to crystal
Purity (Methanolysis Method) min. 98.0 %
Solubility in N,N-DMF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR