3'-(Methylthio)acetanilide

Toimittaja: TCI EUROPE

TCIAM1671-25GEA 198 EUR
TCIAM1671-25G
3'-(Methylthio)acetanilide
3'-(Methylthio)acetanilide
Kaava: C₉H₁₁NOS
CAS-numero: 2524-78-9
https://fi.vwr.com/stibo/web/std.lang.all/59/60/28975960.jpg

Order Now

Testitulosten erittely

Appearance White to Amber to Dark green powder to crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR