4'-Amino-3'-(trifluoromethyl)acetanilide ≥98.0% (by titrimetric analysis)
Toimittaja: TCI EUROPE
TCIAA1895-5GEA
105
EUR
TCIAA1895-5G
4'-Amino-3'-(trifluoromethyl)acetanilide ≥98.0% (by titrimetric analysis)
4'-Amino-3'-(trifluoromethyl)acetanilide
Kaava:
C₉H₉F₃N₂O |
MDL numero:
MFCD00270762 CAS-numero: 1579-89-1 |
Testitulosten erittely
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...