4-Bromo-N-cyclohexylbenzamide, 97%

H57810.03EA 404 EUR
ALFAH57810.03 ALFAH57810.MD
4-Bromo-N-cyclohexylbenzamide, 97%
Purity: 97%
CAS No.: 223553-87-5
Molecular Formula: C13H16BrNO
Formula Weight: 282.18
MDL No.: MFCD00451750
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR