4-Phenoxybenzonitrile ≥98.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAP1649-5GEA
86.9
EUR
TCIAP1649-5G
4-Phenoxybenzonitrile ≥98.0% (by GC)
4-Phenoxybenzonitrile
Kaava:
C₁₃H₉NO MW: 195,22 g/mol Kiehumispiste: 174°C |
MDL numero:
MFCD00017346 CAS-numero: 3096-81-9 ADR: 6.1,III |
Testitulosten erittely
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...