3-Nitrophenylsulphur pentafluoride ≥96.0% (by GC)

Toimittaja: TCI EUROPE
Vaara

TCIAN0742-1GEA 69.3 EUR
TCIAN0742-1G
3-Nitrophenylsulphur pentafluoride ≥96.0% (by GC)
3-Nitrophenylsulphur pentafluoride
Kaava: C₆H₄F₅NO₂S
MW: 249,16 g/mol
Kiehumispiste: 83°C
MDL numero: MFCD00221621
CAS-numero: 2613-26-5
EINECS: 447-600-2
UN: 2810
ADR: 6.1,III
https://fi.vwr.com/stibo/web/std.lang.all/99/56/28989956.jpg

Order Now

Testitulosten erittely

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR