Potassium 4-benzyloxy-2-methylphenyltrifluoroborate, 98%
H52626.03EA 156 EUR
ALFAH52626.03 ALFAH52626.MD
Potassium 4-benzyloxy-2-methylphenyltrifluoroborate, 98%
Purity: 98%
CAS No.: 850623-43-7
Molecular Formula: C14H13BF3KO
Formula Weight: 304.16
Melting Point: 196-200°
MDL No.: MFCD04115740