4-Bromo-N-(3-fluorobenzyl)-2-methylaniline, 97%

H57669.MDEA 75.7 EUR
ALFAH57669.MD ALFAH57669.03
4-Bromo-N-(3-fluorobenzyl)-2-methylaniline, 97%
Purity: 97%
CAS No.: 1019603-22-5
Molecular Formula: C14H13BrFN
Formula Weight: 294.16
MDL No.: MFCD11142044
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR