5-Chloro-N-(4-fluorobenzyl)-2-methylaniline, 97%

H57364.03EA 304 EUR
ALFAH57364.03 ALFAH57364.MD
5-Chloro-N-(4-fluorobenzyl)-2-methylaniline, 97%
Purity: 97%
CAS No.: 1041586-19-9
Molecular Formula: C14H13ClFN
Formula Weight: 249.71
MDL No.: MFCD11121028
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR