4-Ethyl-N-(2-fluorobenzyl)aniline, 97%

H57523.03EA 404 EUR
ALFAH57523.03 ALFAH57523.MD
4-Ethyl-N-(2-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1019570-78-5
Molecular Formula: C15H16FN
Formula Weight: 229.30
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR