5-Fluoro-N-(2-fluorobenzyl)-2-methylaniline, 97%

H57754.MDEA 182 EUR
ALFAH57754.MD ALFAH57754.03
5-Fluoro-N-(2-fluorobenzyl)-2-methylaniline, 97%
Purity: 97%
CAS No.: 1019491-33-8
Molecular Formula: C14H13F2N
Formula Weight: 233.26
MDL No.: MFCD11145961
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR