3-Fluoro-N-(3-fluorobenzyl)-5-methylaniline, 97%

H57198.MDEA 152 EUR
ALFAH57198.MD ALFAH57198.03
3-Fluoro-N-(3-fluorobenzyl)-5-methylaniline, 97%
Purity: 97%
Molecular Formula: C14H13F2N
Formula Weight: 233.26
MDL No.: MFCD22682903
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR