6-Morpholinonicotinic acid

MAYBCC17501DAEA 0 EUR
MAYBCC17501DA MAYBCC17501CB
6-Morpholinonicotinic acid
6-Morpholinonicotinic acid
Kaava: C₁₀H₁₂N₂O₃
MDL numero: MFCD02682034
CAS-numero: 120800-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR