4-Bromo-3-fluoro-N-(4-fluorobenzyl)aniline, 97%

H57335.03EA 280 EUR
ALFAH57335.03 ALFAH57335.MD
4-Bromo-3-fluoro-N-(4-fluorobenzyl)aniline, 97%
Purity: 97%
Molecular Formula: C13H10BrF2N
Formula Weight: 298.13
MDL No.: MFCD21525876
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR