3-Chloro-N-(4-fluorobenzyl)-4-methoxyaniline, 97%
H57249.03EA 304 EUR
ALFAH57249.03 ALFAH57249.MD
3-Chloro-N-(4-fluorobenzyl)-4-methoxyaniline, 97%
Purity: 97%
CAS No.: 861226-54-2
Molecular Formula: C14H13ClFNO
Formula Weight: 265.71