2-Ethoxy-N-(4-fluorobenzyl)aniline, 97%

H57490.03EA 214 EUR
ALFAH57490.03 ALFAH57490.MD
2-Ethoxy-N-(4-fluorobenzyl)aniline, 97%
Purity: 97%
Molecular Formula: C15H16FNO
Formula Weight: 245.29
MDL No.: MFCD11141662
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR