Fast blue B salt zinc double salt, Sigma-Aldrich®

Toimittaja: SIGMA-ALDRICH MICROSCOPY

D9805-100GEA 418 EUR
D9805-100G D9805-10G
Fast blue B salt zinc double salt, Sigma-Aldrich®
Fast blue B salt zinc double salt
Kaava: C₁₄H₁₂Cl₄N₄O₂Zn
MDL numero: MFCD00075203
CAS-numero: 14263-94-6
EINECS: 238-153-2
UN: 0000

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR