Allylcyclohexane ≥97.0% (by GC)

Toimittaja: TCI EUROPE
Vaara

TCIAA1201-5MLEA 47.3 EUR
TCIAA1201-5ML TCIAA1201-25ML
Allylcyclohexane ≥97.0% (by GC)
Allylcyclohexane
Kaava: C₉H₁₆
MW: 124,23 g/mol
Kiehumispiste: 151°C
Leimahduspiste: 29°C
MDL numero: MFCD00013769
CAS-numero: 2114-42-3
EINECS: 218-316-4
UN: 3295
ADR: 3,III
https://fi.vwr.com/stibo/web/std.lang.all/90/60/28929060.jpg

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR