2,6-Dichloropyridine-N-oxide ≥98.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAD3219-5GEA 231 EUR
TCIAD3219-5G
2,6-Dichloropyridine-N-oxide ≥98.0% (by GC)
2,6-Dichloropyridine-N-oxide
Kaava: C₅H₃Cl₂NO
MW: 163,99 g/mol
MDL numero: MFCD00130246
CAS-numero: 2587-00-0
ADR: 6.1,III
https://fi.vwr.com/stibo/web/std.lang.all/02/09/28980209.jpg

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 138.0 to 142.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR