2,1,3-Benzothiadiazole-4-sulphonyl chloride

MAYBCC09103DAEA 0 EUR
MAYBCC09103DA MAYBCC09103.250
2,1,3-Benzothiadiazole-4-sulphonyl chloride
2,1,3-Benzothiadiazole-4-sulphonyl chloride
Kaava: C₆H₃ClN₂O₂S₂
MDL numero: MFCD00068049
CAS-numero: 73713-79-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR