2-Isobutyl-4-methylthiazole ≥96.0% (by GC)
Toimittaja: TCI EUROPE
TCIAI0442-5MLEA
96.8
EUR
TCIAI0442-5ML
2-Isobutyl-4-methylthiazole ≥96.0% (by GC)
2-Isobutyl-4-methylthiazole
Kaava:
C₈H₁₃NS MW: 155,2605 g/mol Kiehumispiste: 189°C |
MDL numero:
MFCD00053138 CAS-numero: 61323-24-8 EINECS: 262-709-3 |
Testitulosten erittely
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...