Hydrogen sulphide cylinder, emptied/unwashed
Vaara
VWRC600583B.EMPTYEA 0 EUR
VWRC600583B.EMPTY
Hydrogen sulphide cylinder, emptied/unwashed
Hydrogen sulphide cylinder, emptied/unwashed
Kaava: H₂S
CAS-numero: 7783-06-4
EINECS: 231-977-3
UN: 1053
ADR: 2,
Merck Index: 12,04843