4-Methoxythioanisole ≥98.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAM1852-5GEA 44 EUR
TCIAM1852-5G TCIAM1852-25G
4-Methoxythioanisole ≥98.0% (by GC)
4-Methoxythioanisole
Kaava: C₈H₁₀OS
Kiehumispiste: 137°C
MDL numero: MFCD00010587
CAS-numero: 1879-16-9
https://fi.vwr.com/stibo/web/std.lang.all/62/74/28976274.jpg

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR