1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane ≥98.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAP1224-10GEA
147
EUR
TCIAP1224-10G
1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane ≥98.0% (by GC)
1,1,1,2,2,3,3-Heptafluoro-3-[(trifluorovinyl)oxy]propane
Kaava:
C₅F₁₀O MW: 266,04 g/mol Kiehumispiste: 36°C Storage Temperature: Jääkaappi |
MDL numero:
MFCD00191577 CAS-numero: 1623-05-8 EINECS: 216-600-2 |
Testitulosten erittely
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...