Bis(triphenylphosphine)copper(I) borohydride ≥90.0% (by titrimetric analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAB1139-5GEA 26.4 EUR
TCIAB1139-5G
Bis(triphenylphosphine)copper(I) borohydride ≥90.0% (by titrimetric analysis)
Bis(triphenylphosphine)copper(I) borohydride
Kaava: C₃₆H₃₄BCuP₂
MW: 602,97 g/mol
MDL numero: MFCD00013219
CAS-numero: 16903-61-0
EINECS: 240-951-0
ADR: 4.3,III
https://fi.vwr.com/stibo/web/std.lang.all/29/84/28932984.jpg

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR