Hexyltrichlorosilane ≥97.0% (by GC)

Toimittaja: TCI EUROPE
Vaara

TCIAH0547-25MLEA 49.5 EUR
TCIAH0547-25ML
Hexyltrichlorosilane ≥97.0% (by GC)
Hexyltrichlorosilane
Kaava: C₆H₁₃Cl₃Si
MW: 219,61 g/mol
Kiehumispiste: 192°C
Leimahduspiste: 56°C
MDL numero: MFCD00013606
CAS-numero: 928-65-4
EINECS: 213-178-1
UN: 1784
ADR: 8,II
https://fi.vwr.com/stibo/web/std.lang.all/05/58/28970558.jpg

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR