(±)-γ-Octalactone ≥96.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAH0232-25GEA
50.6
EUR
TCIAH0232-25G
(±)-γ-Octalactone ≥96.0% (by GC)
(±)-γ-Octalactone
Kaava:
C₈H₁₄O₂ MW: 142,2 g/mol Kiehumispiste: 133°C Leimahduspiste: 126°C |
MDL numero:
MFCD00005402 CAS-numero: 104-50-7 EINECS: 203-208-1 |
Testitulosten erittely
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...