(±)-δ-Hexalactone ≥99.0% (by GC)
Toimittaja: TCI EUROPE
TCIAH0759-25GEA
45.1
EUR
TCIAH0759-25G
(±)-δ-Hexalactone ≥99.0% (by GC)
(±)-δ-Hexalactone
Kaava:
C₆H₁₀O₂ MW: 114,14 g/mol Kiehumispiste: 115°C Leimahduspiste: 103°C |
MDL numero:
MFCD00083574 CAS-numero: 823-22-3 EINECS: 212-511-8 |
Testitulosten erittely
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...