(±)-δ-Nonalactone ≥98.0% (by GC)
Toimittaja: TCI EUROPE
TCIAN0424-25MLEA
34.1
EUR
TCIAN0424-25ML
(±)-δ-Nonalactone ≥98.0% (by GC)
(±)-δ-Nonalactone
Kaava:
C₉H₁₆O₂ MW: 156,22 g/mol Kiehumispiste: 137°C Leimahduspiste: 137°C |
MDL numero:
MFCD00036500 CAS-numero: 3301-94-8 EINECS: 221-974-5 |
Testitulosten erittely
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...