(±)-Pipecolinic acid ≥98.0% (by titrimetric analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAP0442-25GEA 49.5 EUR
TCIAP0442-25G
(±)-Pipecolinic acid ≥98.0% (by titrimetric analysis)
(±)-Pipecolinic acid
Kaava: C₆H₁₁NO₂
MW: 129,16 g/mol
MDL numero: MFCD00064347
CAS-numero: 535-75-1
EINECS: 208-616-3
https://fi.vwr.com/stibo/web/std.lang.all/11/31/28991131.jpg

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR