Osmiumtetraoksidi 4% in water

Toimittaja: TCI EUROPE
Vaara

TCIAO0308-10MLEA 136 EUR
TCIAO0308-10ML
Osmiumtetraoksidi 4% in water
Osmiumtetraoksidi
Kaava: O₄Os
MW: 254,23 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00011150
CAS-numero: 20816-12-0
EINECS: 244-058-7
UN: 3289
ADR: 6.1,II
https://fi.vwr.com/stibo/web/std.lang.all/05/71/28990571.jpg

Order Now

Testitulosten erittely

Appearance Light orange to Yellow to Green clear liquid
Concentration 4.0 to 4.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR