1-Bromo-5-fluoro-2-nitrobenzene ≥98.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAB3061-5GEA 64.9 EUR
TCIAB3061-5G
1-Bromo-5-fluoro-2-nitrobenzene ≥98.0% (by GC)
1-Bromo-5-fluoro-2-nitrobenzene
Kaava: C₆H₃BrFNO₂
MW: 220 g/mol
MDL numero: MFCD00792441
CAS-numero: 700-36-7
https://fi.vwr.com/stibo/web/std.lang.all/45/03/28934503.jpg

Order Now

Testitulosten erittely

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR