Triisopentylamine ≥95.0% (by GC)
Toimittaja: TCI EUROPE
|
Vaara
|
TCIAT0455-10MLEA
29.7
EUR
TCIAT0455-10ML
Triisopentylamine ≥95.0% (by GC)
Triisopentylamine
Kaava:
C₁₅H₃₃N MW: 227,43 g/mol Kiehumispiste: 270°C Leimahduspiste: 77°C |
MDL numero:
MFCD00015054 CAS-numero: 645-41-0 EINECS: 211-441-5 UN: 2735 ADR: 8,III |
Testitulosten erittely
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...