6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedione ≥97.0% (by GC)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAD1729-5GEA
77
EUR
TCIAD1729-5G
6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedione ≥97.0% (by GC)
6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedione
Kaava:
C₁₀H₁₁F₇O₂ MW: 296,18 g/mol Sulamispiste: 34…35 °C Leimahduspiste: 53°C |
MDL numero:
MFCD00000435 CAS-numero: 17587-22-3 EINECS: 241-556-6 UN: 1993 ADR: 3,III |
![]() |
Testitulosten erittely
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...