Methyl-4-iodo-2-methoxybenzoate ≥98.0% (by GC)

Toimittaja: TCI EUROPE

TCIAM2096-5GEA 52.8 EUR
TCIAM2096-5G TCIAM2096-25G
Methyl-4-iodo-2-methoxybenzoate ≥98.0% (by GC)
Methyl-4-iodo-2-methoxybenzoate
Kaava: C₉H₉IO₃
MW: 292,07 g/mol
MDL numero: MFCD00052925
CAS-numero: 35387-93-0
https://fi.vwr.com/stibo/web/std.lang.all/80/53/28988053.jpg

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR