2-Klooribentsoehappo, zone refined: number of passes 20

Toimittaja: TCI EUROPE
Varoitus

TCIAC0726-ZONEEA 113 EUR
TCIAC0726-ZONE
2-Klooribentsoehappo, zone refined: number of passes 20
2-Klooribentsoehappo
Kaava: C₇H₅ClO₂
MW: 156,57 g/mol
Sulamispiste: 139…142 °C
MDL numero: MFCD00002412
CAS-numero: 118-91-2
EINECS: 204-285-4
Merck Index: 13,02143
https://fi.vwr.com/stibo/web/std.lang.all/77/72/28937772.jpg

Order Now

Testitulosten erittely

Appearance White powder to crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR