5-(Trimethylsilyl)thiazole ≥98.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAT1523-1MLEA 68.2 EUR
TCIAT1523-1ML
5-(Trimethylsilyl)thiazole ≥98.0% (by GC)
5-(Trimethylsilyl)thiazole
Kaava: C₆H₁₁NSSi
MW: 157,31 g/mol
Kiehumispiste: 65°C
MDL numero: MFCD00191506
CAS-numero: 79265-36-4
https://fi.vwr.com/stibo/web/std.lang.all/55/37/28995537.jpg

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR