4-Isopropylbenzoic acid ≥98.0% (by GC, titration analysis)
Toimittaja: TCI EUROPE
|
Varoitus
|
TCIAI0169-25GEA
108
EUR
TCIAI0169-25G
4-Isopropylbenzoic acid ≥98.0% (by GC, titration analysis)
4-Isopropylbenzoic acid
Kaava:
C₁₀H₁₂O₂ MW: 164,2 g/mol Kiehumispiste: 125°C Sulamispiste: 115…117 °C |
MDL numero:
MFCD00002564 CAS-numero: 536-66-3 EINECS: 208-642-5 Merck Index: 12,02684 |
![]() |
Testitulosten erittely
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 116.0 to 120.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...