4-Pentylbenzoic acid ≥98.0% (by titrimetric analysis)

Toimittaja: TCI EUROPE
Varoitus

TCIAA0741-250GEA 362 EUR
TCIAA0741-250G TCIAA0741-25G TCIAA0741-5G
4-Pentylbenzoic acid ≥98.0% (by titrimetric analysis)
4-Pentylbenzoic acid
Kaava: C₁₂H₁₆O₂
MW: 192,26 g/mol
Sulamispiste: 85…88 °C
MDL numero: MFCD00002572
CAS-numero: 26311-45-5
EINECS: 247-607-9
https://fi.vwr.com/stibo/web/std.lang.all/84/30/28928430.jpg

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 86.0 to 91.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR