Deoxybenzoin ≥98.0% (by GC)
Toimittaja: TCI EUROPE
TCIAB0435-250GEA 130 EUR
TCIAB0435-250G TCIAB0435-25G
Deoxybenzoin ≥98.0% (by GC)
Deoxybenzoin
Kaava: C₁₄H₁₂O
MW: 196,25 g/mol
Kiehumispiste: 322°C
Storage Temperature: Jääkaappi
MDL numero: MFCD00003081
CAS-numero: 451-40-1
EINECS: 207-193-2
https://fi.vwr.com/stibo/web/std.lang.all/24/02/28932402.jpg
Order Now
Testitulosten erittely
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C
Solubility in Methanol almost transparency