3-Methylthiophene-2-carbonyl chloride ≥98.0% (by GC, titration analysis)

Toimittaja: TCI EUROPE
Vaara

TCIAM1711-5GEA 114 EUR
TCIAM1711-5G
3-Methylthiophene-2-carbonyl chloride ≥98.0% (by GC, titration analysis)
3-Methylthiophene-2-carbonyl chloride
Kaava: C₆H₅ClOS
MW: 160,62 g/mol
Kiehumispiste: 220°C
MDL numero: MFCD00130091
CAS-numero: 61341-26-2
UN: 3265
ADR: 8,II
https://fi.vwr.com/stibo/web/std.lang.all/60/27/28976027.jpg

Order Now

Testitulosten erittely

Appearance White to Almost white clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR