Methyl-3,5-dibromo-4-hydroxybenzoate ≥99.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAD2501-25GEA 52.8 EUR
TCIAD2501-25G
Methyl-3,5-dibromo-4-hydroxybenzoate ≥99.0% (by GC)
Methyl-3,5-dibromo-4-hydroxybenzoate
Kaava: C₈H₆Br₂O₃
MW: 309,94 g/mol
MDL numero: MFCD00016416
CAS-numero: 41727-47-3
EINECS: 255-521-8
https://fi.vwr.com/stibo/web/std.lang.all/94/28/28979428.jpg

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 122.0 to 125.0 deg-C
Solubility in Methanol within very faint turbidity
Water 5.0 to 6.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR