2-Deoxy-2-fluoro-D-glucose ≥98.0% (by GC)

Toimittaja: TCI EUROPE

TCIAD3023-100MGEA 347 EUR
TCIAD3023-100MG
2-Deoxy-2-fluoro-D-glucose ≥98.0% (by GC)
2-Deoxy-2-fluoro-D-glucose
Kaava: C₆H₁₁FO₅
Storage Temperature: Pakastin
MDL numero: MFCD00077527
CAS-numero: 29702-43-0
https://fi.vwr.com/stibo/web/std.lang.all/01/43/28980143.jpg

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +60.0 to +65.0 deg (C=1,dil ammonia water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR