Naphthalene black 10B (naphthol blue black, acid black 1) ≥95.0% (by HPLC) elektroforeesiin
Toimittaja: TCI EUROPE
Varoitus
TCIAA2097-5GEA 17.6 EUR
TCIAA2097-5G
Naphthalene black 10B (naphthol blue black, acid black 1) ≥95.0% (by HPLC) elektroforeesiin
Naphthalene black 10B (naphthol blue black, acid black 1)
Kaava: C₂₂H₁₄N₆Na₂O₉S₂
MW: 616,49 g/mol
MDL numero: MFCD00004017
CAS-numero: 1064-48-8
EINECS: 213-903-1
https://fi.vwr.com/stibo/web/std.lang.all/02/53/28930253.jpg
Order Now
Testitulosten erittely
Appearance Light yellow to Amber to Dark green powder to crystaline
Purity (HPLC) min. 95.0 area%
Lambda max. 616.0 to 620.0 nm (H2O)
Absorbance (E1%1cm) min. 700 (H2O, 616.0 to 620.0 nm)
Functionality test to pass test