8-Octanelactam ≥98.0% (by GC)

Toimittaja: TCI EUROPE
Varoitus

TCIAO0162-10GEA 120 EUR
TCIAO0162-10G
8-Octanelactam ≥98.0% (by GC)
8-Octanelactam
Kaava: C₈H₁₅NO
MW: 141,21
Kiehumispiste: 151°C
MDL numero: MFCD00003273
CAS-numero: 935-30-8
EINECS: 213-300-3
https://fi.vwr.com/stibo/web/std.lang.all/04/54/28990454.jpg

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 78.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR